Barrel valve
Catalog # BURK0536-0010
Supplier: BURKLE
Specifications
- For:metal barrel with 2” BSP TriSure metal thread
- Environmentally Preferable:
- Pk:1
Specifications
About this item
The barrel valve for opening the barrel closure as well as emptying and reclosing it even with lying, full barrels without having to move them. Also used for highly viscous liquids.