Multi-layer adhesive floor mats for cleanrooms, CleanStep™
Supplier: Texwipe
Certificates
About this item
Adhesive coated contamination control mats capture particulates from foot traffic and equipment wheels, providing the most effective dirt and dust removal method available in a mat.
- Self-adhesive base prevents slipping
- Consistent adhesive coating on each layer
- Anti-microbial; reduces the potential of spreading germs and bacteria
- Low profile (approx. 1,5 mm for 30 layers, 2 mm for 60 layers) prevents tripping
- Each layer is numbered, so the number of layers remaining is easily identified
- Clear film layers over colour bottom film for colour consistency through life of product
- Available in 30 and 60 layers
Colour: Blue, grey or white
Frequently Bought Together