Ampoules, pre-scored, WHEATON®
Supplier: DWK Life Sciences
Certificates
About this item
Glass ampoules made from low extractable borosilicate glass can be hermetically sealed to preserve purity and extend content shelf life.
- Tamper-evident seal
- For biological product preservation
- Gold band on the stem of the ampoule indicates that it is pre-scored to facilitate snapping off the top eliminating the need to file the ampoule
Ideal for standards packaging, environmental and diagnostic standards, lyophilisation and cryogenic storage.
Certifications: USP Type I, ASTM E438 Type I, Class A
Frequently Bought Together