(2-Hydroxyethyl)hydrazine
: β-Hydroxyethylhydrazine
:109-84-2
:
:C₂H₈N₂O
:MFCD00007623
:218…220 °C
:–70 °C
:1.121
:76.1 g/mol
:73 °C (163 °F)
:Ambient
:203-711-6
:2810
:6.1 II
:GHS06
:GHS07
- Environmentally Preferable:
Frequently Bought Together