Order Entry
Italy
ContactUsLinkComponent
Corning® Dispase (from Bacillus polymyxa)
Corning® Dispase (from Bacillus polymyxa)
  734-1312
 :  Corning
   Low Manufacturing Impact
 :  
Corning® Dispase (from Bacillus polymyxa)
  734-1312
 :  Corning
   Low Manufacturing Impact
 :  

 

  • Pk:
    100 mL
  • Source:
    Bacillus polymyxa
  • Species:
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Storage conditions:
    Stable for at least three months at −20 °C.
  • Environmental certification:
    ISO 14001 - Bedford
  • Enzyme name:
    Dispase
  • Enzyme activity:
    5000 CU
  • Formulation:
    Frozen in HBSS, pH 7.4

 

 

Dispase is a bacillus-derived neutral metalloprotease that is recommended for recovering cell cultured on Corning® Matrigel® Basement Membrane Matrix. Dispase will yield a single cell suspension far more gently and effectively than trypsin, collagenase, or other proteolytic enzymes; it will not harm cells harvested for sub cultivation or bioassays. In addition, Dispase may be used for tissue dissociation. Dispase cleaves fibronectin, collagen IV, and to a lesser extent collagen I, but is does not cleave collagen V or laminin.

  • Tested for ability to solubilise a 1 mm thick layer of gelled BD Matrigel™ matrix after two hours at 37 °C
  • Filtered and tested and found negative for bacteria, fungi, and mycoplasma