4-Phenylpyridine 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:939-23-1
:
Formula:C₁₁H₉N
MDL Number:MFCD00006420
Boiling Pt:274…275 °C (760 mmHg)
Molecular Weight:155.2 g/mol
Melting Pt:71…75 °C
EINECS:213-357-4
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to light yellow to beige Powder or crystals
- Infrared spectrum:Conforms
- Melting point:71 to 75 °C
- GC:≥98.5 %
- Residue after evaporation:≤0.001 %
Specification Test Results
Frequently Bought Together