Naphthalene 99%
:91-20-3
:
:C₁₀H₈
:MFCD00001742
:218 °C (760 mmHg)
:79…82 °C
:0.99 g/cm³
:128.17 g/mol
:78 °C
:1334
:4.1 III
:12,06457
:202-049-5
:GHS02
:GHS07
:GHS08
:GHS09
:Ambient
- Environmentally Preferable:
- Appearance:White to off-white Crystalline powder or crystals or flakes or tablets or balls
- Infrared spectrum:Conforms
- Melting point:79 to 82 °C
- GC:≥98.5 %
Frequently Bought Together