Water, gradient grade for HPLC
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Hydrogen oxide
CAS Number:7732-18-5
:
Formula:H₂O
MDL Number:MFCD00011332
Boiling Pt:100
Melting Pt:0
Density:1 g/cm³
Molecular Weight:18.02 g/mol
Merck Index:13,10098
Storage Temperature:Ambient
EINECS:231-791-2
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Clear colorless Liquid
- Residue after evaporation:≤0.0005 %
- Residue after ignition:≤0.0003 %
- Gradient analysis::
- Absorbance largest eluted peak:≤0.002 A.U.
- Filtered to 0.2 micrometer:Filtered
Specification Test Results
Frequently Bought Together