Order Entry
Italy
ContactUsLinkComponent
 
Sodium azide 99%, extra pure
 
undefined
Sodium azide 99%, extra pure
 

 

  • Environmentally Preferable:

 

 

  • Appearance:
    White Crystalline powder
  • Assay:
    ≥98.5 %
  • Loss on drying:
    ≤0.5 %
  • pH:
    ≥9 (5 % solution)
  • Sodium carbonate (Na2CO3):
    ≤0.15 %
  • Sodium hydroxide (NaOH):
    ≤0.1 %
  • Heavy metals (as Pb):
    ≤20 ppm
  • Insoluble matter:
    ≤0.05 % (in water)

 

Frequently Bought Together