Order Entry
Italy
ContactUsLinkComponent
Sodium bromate ≥99%, Fluka™
Sodium bromate ≥99%, Fluka™
Certificates
Sodium bromate ≥99%, Fluka™

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Assay:
    Min. 99 %
  • Iron (Fe):
    Max. 0.005 %
  • Heavy metals (as Pb):
    Max. 0.005 %
  • Bromide (Br):
    Max. 0.05 %
  • Sulfate (SO₄):
    Max. 0.05 %

Specification Test Results

Frequently Bought Together