Order Entry
Italy
ContactUsLinkComponent
(S)-(-)-α-Terpineol, Sigma-Aldrich®
(S)-(-)-α-Terpineol, Sigma-Aldrich®
Supplier:  Merck
Certificates
(S)-(-)-α-Terpineol, Sigma-Aldrich®
Supplier:  Merck
Certificates

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Solubility:
    (20 °C) insoluble
  • Melting Point:
    32 - 34 °C
  • Molar Mass:
    154.25 g/mol
  • Boiling Point:
    215 - 217 °C (1013 hPa)
  • Flash Point:
    90 °C
  • Refractive Index:
    1.4813 (20 °C, 589 nm)
  • Density:
    0.935 g/cm3 (20 °C)

Specification Test Results

Related Information

Frequently Bought Together