N-Methylmorpholine-N-oxide 50% (w/w) in aqueous solution
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: 4-Methylmorpholine N-Oxide, 4-Methylmorpholine 4-oxide
CAS Number:7529-22-8
:
Formula:C₅H₁₁NO₂
MDL Number:MFCD00005947
Boiling Pt:118…119 °C
Melting Pt:–20 °C
Density:1.140
Molecular Weight:117.15 g/mol
Storage Temperature:Refrigerator
EINECS:231-391-8
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together