Ethylbenzene
Catalog # PROOCHEUSC12
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:2 mL
- Seq:2145
- Environmentally Preferable:
Specifications
About this item
Synonyms: Phenylethane
:
:106.17 g/mol
Melting Pt:–95 °C
EINECS:202-849-4
UN:1175
:GHS02
:GHS07
Storage Temperature:Ambient
CAS Number:100-41-4
Formula:C₈H₁₀
MDL Number:MFCD00011647
Boiling Pt:136.2
Density:0.867 g/cm³ (20 °C)
Flash Pt:15
ADR:3 II
Merck Index:13,03801