Benzyl chloride for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: (Chloromethyl)benzene, Chlorophenylmethane, α-Chlorotoluene
CAS Number:100-44-7
:
Formula:(C₆H₅)CH₂Cl
MDL Number:MFCD00000889
Boiling Pt:179.3
Melting Pt:–41.2 °C
Density:1.103 g/cm³ (20 °C)
Molecular Weight:126.59 g/mol
Flash Pt:52
EINECS:202-853-6
UN:1738
ADR:6.1 (8) II
Merck Index:13,01131
:M-Clarity™ quality level = MQ200
:GHS06
:GHS05
:GHS08
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Solubility:460 mg/l (30 °C)
- Melting Point:-41.2 °C
- Molar Mass:126.58 g/mol
- Boiling Point:179.3 °C (1013 hPa)
- Vapor Pressure:1.2 hPa (20 °C)
- Flash Point:67 °C
- Refractive Index:1.5415
- Explosion Limit:1.1 - 14 %(V)
- Density:1.10 g/cm3 (20 °C)
- pH:(H2O) not applicable
- Ignition Point:585 °C
- Hazard Class:6.1
- Hazard Class Description:Toxic materials
Specification Test Results
Related Information
Certificates
Search by Lot Numbermsds_801809
SDS
ProductPage_801809 ProductPage_801809
Product specification
Frequently Bought Together