Acetophenone 98%, pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Methyl phenyl ketone, Phenyl methyl ketone
CAS Number:98-86-2
:
Formula:C₈H₈O
MDL Number:MFCD00008724
Boiling Pt:202 °C (760 mmHg)
Melting Pt:19.6 °C
Density:1.028 g/cm³
Molecular Weight:120.15 g/mol
Flash Pt:77 °C
Merck Index:13,00074
EINECS:202-708-7
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5320 to 1.5350 (20°C, 589 nm)
- GC:≥97.5 %
- Visual transition interval:from pH 3.5 (yellow) to pH 6.5 (pink)
- BSC Certification:Passes
Specification Test Results
Frequently Bought Together