Phenol 99%, extra pure
: Phenylalcohol, Carbolic acid, Hydroxybenzene
:108-95-2
:
:C₆H₅OH
:MFCD00002143
:182 °C (760 mmHg)
:39…42 °C
:1.07 g/cm³
:94.11 g/mol
:79 °C
:Ambient
:1671
:6.1 II
:12,07390
:203-632-7
:01-2119471329-32
:GHS05
:GHS06
:GHS08
:GHS09
- Environmentally Preferable:
- Appearance:Colorless to white Crystals or fused mass
- Infrared spectrum:Conforms
- Melting point:39 to 42 °C
- Water:≤0.2 %
- GC:≥98.5 %
Frequently Bought Together