6-Methyl-2-heptanone ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
Synonyms: Methyl 4-Methylpentyl ketone, 6-Methylheptan-2-one
CAS Number:928-68-7
:
Formula:C₈H₁₆O
MDL Number:MFCD00026527
Boiling Pt:171°C
Molecular Weight:128.21 g/mol
Melting Pt:-59
EINECS:213-179-7
UN:1224
:GHS02
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 98.0 %
- Acid Value:max. 3.0
Specification Test Results
Frequently Bought Together