3-Nitroaniline
Catalog # EHERC15554300
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,25 g
- Seq:3029
- Environmentally Preferable:
Specifications
About this item
Synonyms: m-Nitroaniline
:
:138.13 g/mol
Melting Pt:109…112 °C
EINECS:202-729-1
UN:1661
:GHS06
:GHS08
Storage Temperature:Ambient
CAS Number:99-09-2
Formula:C₆H₆N₂O₂
MDL Number:MFCD00007782
Boiling Pt:306
Density:0.9 g/cm³ (25 °C)
Flash Pt:196
ADR:6.1 II
Merck Index:12,06679