4-Hydroxybenzoic acid 99+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: p-Hydroxybenzoic Acid
CAS Number:99-96-7
:
Formula:C₇H₆O₃
MDL Number:MFCD00002547
Boiling Pt:336
Molecular Weight:138.12 g/mol
Melting Pt:214…217 °C
Flash Pt:199 °C
Merck Index:13,04837
EINECS:202-804-9
:GHS05
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to cream Crystalline powder
- Infrared spectrum:Conforms
- Melting point:214 to 217 °C
- HPLC:≥99.0 %
- Titration with NaOH:≥99.0 %
Specification Test Results
Frequently Bought Together