3,5-Dinitrobenzoic acid 98+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:99-34-3
:
Formula:C₇H₄N₂O₆
MDL Number:MFCD00007253
Density:1.683 g/cm³
Molecular Weight:212.12 g/mol
Melting Pt:205…209 °C
Merck Index:13,03304
EINECS:202-751-1
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to yellow Crystalline powder or crystals
- Infrared spectrum:Conforms
- Melting point:204 to 209 °C
- HPLC:≥98.0 %
- Titration with NaOH:98.0 to 102.0 %
Specification Test Results
Frequently Bought Together