Benzoic acid methyl ester for synthesis, Sigma-Aldrich®
: Merck
: Methyl benzoate
:93-58-3
:
:C₆H₅COOCH₃
:MFCD00008421
:199.6
:–12 °C
:1.0888 g/cm³ (20 °C)
:136.15 g/mol
:83
:13,06052
:M-Clarity™ quality level = MQ200
:202-259-7
:GHS07
:Ambient
- Environmentally Preferable:
- Solubility:0.157 g/l (30 °C)
- Melting Point:-12 °C
- Molar Mass:136.15 g/mol
- Boiling Point:199 °C (1013 hPa)
- Vapor Pressure:0.36 hPa (20 °C)
- Flash Point:82 °C
- Explosion Limit:8.6 - 20 %(V)
- Density:1.09 g/cm3 (20 °C)
- Ignition Point:510 °C
- Hill Formula:C8H8O2
- Chemical Formula:C6H5COOCH3
Frequently Bought Together