Sodium dihydrogen citrate, anhydrous 99%, pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Sodium 3,4-dicarboxy-3-hydroxybutanoate, Sodium citrate monobasic, mono-Sodium citrate, Citric acid monosodium salt, Monosodium Citrate
CAS Number:18996-35-5
:
Formula:C₆H₇NaO₇
MDL Number:MFCD00013067
Molecular Weight:214.11 g/mol
Melting Pt:209…215 °C
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Titration with HClO4:≥98.5 %
Specification Test Results
Frequently Bought Together