1-Bromo-3,5-dichlorobenzene 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: 3,5-Dichlorobromobenzene
CAS Number:19752-55-7
:
Formula:C₆H₃BrCl₂
MDL Number:MFCD00000584
Boiling Pt:232 °C
Molecular Weight:225.9 g/mol
Melting Pt:74…77 °C
Flash Pt:>110 °C
EINECS:243-270-7
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to beige to light brown Crystalline powder
- Infrared spectrum:Conforms
- Melting point:74 to 77 °C
- GC:≥97.5 %
Specification Test Results
Frequently Bought Together