1-Chloro-2-methylpropane ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:513-36-0
:
Formula:C₄H₉Cl
MDL Number:MFCD00000954
Boiling Pt:68…69 °C
Melting Pt:–131 °C
Density:0.879
Molecular Weight:92.57 g/mol
Flash Pt:–7 °C (19 °F)
UN:1127
ADR:3 II
Merck Index:13,05155
EINECS:208-157-9
:GHS02
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together