1,4-Butanediol 99+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: 1,4-Dihydroxy butane, BDO, Tetramethylene glycol, 1,4-Butylene glycol
CAS Number:110-63-4
:
Formula:C₄H₁₀O₂
MDL Number:MFCD00002968
Boiling Pt:229.2 °C (1013hPa)
Melting Pt:20 °C
Density:1.01 g/cm³
Molecular Weight:90.12 g/mol
Flash Pt:135 °C
EINECS:203-786-5
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Clear viscous liquid or low melting solid
- Infrared spectrum:Conforms
- Refractive index:1.4440 to 1.4470 (20°C, 589 nm)
- Water:≤0.1 % (K.F.)
- GC:≥99.0 %
- Color scale:≤10 APHA
Specification Test Results
Frequently Bought Together