2-Chlorobutane 99+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:78-86-4
:
Formula:C₄H₉Cl
MDL Number:MFCD00000871
Boiling Pt:68
Melting Pt:–140 °C
Density:0.87 g/cm³
Molecular Weight:92.57 g/mol
Flash Pt:–15 °C
UN:1127
ADR:3 II
Merck Index:13,01560
EINECS:201-151-7
:GHS02
:GHS05
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.3960 to 1.3980 (20°C, 589 nm)
- GC:≥99.0 %
- UV:at 219 nm A: ≤1
- Color scale:≤10 APHA
- Residue after evaporation:≤0.0005 %
Specification Test Results
Frequently Bought Together