Capric acid
Catalog # EHERC12095050
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,25 g
- Seq:3029
- Environmentally Preferable:
Specifications
About this item
Synonyms: Decylic acid, Decanoic acid
:
:172.27 g/mol
Boiling Pt:268
EINECS:206-376-4
:GHS07
Melting Pt:29…32 °C
Storage Temperature:Ambient
CAS Number:334-48-5
Formula:C₁₀H₂₀O₂
MDL Number:MFCD00004441
Density:0.893 g/cm³ (20 °C)
Flash Pt:147
Merck Index:13,01764