Acetylacetone for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: ACAC, 2,4-Pentanedione
CAS Number:123-54-6
:
Formula:CH₃COCH₂COCH₃
MDL Number:MFCD00008787
Boiling Pt:140.4
Melting Pt:–23 °C
Density:0.96945 g/cm³ (20 °C)
Molecular Weight:100.12 g/mol
Flash Pt:30
EINECS:204-634-0
UN:2310
ADR:3 (6.1) III
Merck Index:13,00083
:M-Clarity™ quality level = MQ200
:GHS02
:GHS06
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Solubility:200 g/l (20 °C)
- Melting Point:-23 °C
- Molar Mass:100.12 g/mol
- Boiling Point:138 - 140 °C (1013 hPa)
- Vapor Pressure:9 hPa (20 °C)
- Flash Point:34 °C
- Explosion Limit:2.4 - 11.4 %(V)
- Density:0.97 g/cm3 (20 °C)
- pH:6 (200 g/l, H2O, 20 °C)
- Ignition Point:340 °C
- Evaporation Number:20
Specification Test Results
Related Information
Frequently Bought Together