Acetaldehyde ≥99%, TECHNICAL
Catalog # 20877.265
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:500 mL
- Seq:2725
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay:Min. 99 %
- Refractive index (20°C):1.330 - 1.332
Specification Test Results
About this item
Synonyms: Ethanal
:
:44.05 g/mol
Flash Pt:–27 °C
EINECS:200-836-8
UN:1089
:GHS02
:GHS08
:GHS07
Storage Temperature:Refrigerator
CAS Number:75-07-0
Formula:CH₃CHO
MDL Number:MFCD00006991
Boiling Pt:20.1 °C (1013 hPa)
Melting Pt:–123 °C
Density:0.78 g/cm³ (20 °C)
ADR:3 I
Merck Index:12,00037