Order Entry
Italy
ContactUsLinkComponent
 
tri-Sodium phosphate dodecahydrate, EMPROVE® ESSENTIAL E339, NF suitable for use as excipient, SAFC®
 
tri-Sodium phosphate dodecahydrate, EMPROVE® ESSENTIAL E339, NF suitable for use as excipient, SAFC®
 

 

  • Environmentally Preferable:

 

 

  • Identity:
    passes test
  • Assay (alkalimetric, calculated on calcined substance):
    ≥ 93.5 %
  • Free alkali (as NaOH):
    ≤ 2.5 %
  • In water insoluble matter (calculated on dried substance):
    ≤ 0.2 %
  • pH-value (1 %, water):
    11.5 - 12.5
  • Fluoride (F):
    ≤ 0.0010 %
  • Heavy metals (as Pb):
    ≤ 0.0010 %
  • As (Arsenic):
    ≤ 0.00005 %
  • Cd (Cadmium):
    ≤ 0.0001 %
  • Hg (Mercury):
    ≤ 0.0001 %
  • Pb (Lead):
    ≤ 0.0001 %
  • Loss on ignition (800 °C):
    45.0 - 57.0 %
  • Residual solvents (ICH Q3C):
    excluded by production process

 

Frequently Bought Together