Iodine ≥99.9985% (metals basis), crystals, Puratronic®, resublimed
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7553-56-2
:
Formula:I₂
MDL Number:MFCD00011355
Boiling Pt:184.3
Melting Pt:183…186 °C (subl.)
Density:4.930
Molecular Weight:253.81 g/mol
Storage Temperature:Ambient
EINECS:231-442-4
ADR:8 III
Merck Index:13,05036
UN:3495
:GHS07
:GHS09
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance (Colour):Grey
- Form:Crystalline
- Total Metal Impurities:≤0.0015% (15 ppm)
Specification Test Results
Frequently Bought Together