Microplate dispensers, Multidrop® Combi
Based on the proven and reliable Multidrop™ range, the Multidrop™ Combi provides high speed bulk reagent dispensing for biotech and pharmaceutical laboratories. The Multidrop™ Combi is remarkably flexible, dispensing into plates ranging from 6-well to 1536-wells with automated dispensing height adjustment for low profile to deep well plates.
- Precise dispensing with peristaltic pump technology across a wide range: 0,5 to 2500 µl
- Reduced reagent costs with low volume dispensing and minimal dead volumes
- Simple operation with icon-based graphic display
- High throughput operation with full robot compatibility
- The Multidrop™ Combi equipped with the SMART feature counts the usage of the dispensing cassette and displays the calibration status
The Multidrop™ Combi combines the most versatile features for reagent dispensing with excellent performance for drug discovery or genomic and proteomic assays.
- Catalog No:
- 735-0394
- 735-0393
- Plate types:
- 6-well to 1536-well plates
- 6-well to 1536-well plates
- Interfaces:
- Onboard software and serial RS-232 or USB for PC control
- Onboard software and serial RS-232 or USB for PC control
- Dispensing volume range:
- Small tube dispensing cassette: 0,5 - 50 µl, in 0,5 µl increments
Standard tube dispensing cassette: 5 - 2500 µl, in 5 µl increments - Small tube dispensing cassette: 0,5 - 50 µl, in 0,5 µl increments
Standard tube dispensing cassette: 5 - 2500 µl, in 5 µl increments
- Dispensing speed:
- 96-well plate: 10 µl in 5 s, 20 µl in 5 s, 100 µl in 10 s
384-well plate: 1 µl in 6 s, 5 µl in 9 s, 10 µl in 13 s, 20 µl in 21 s
1536-well plate: 1 µl in 15 s, 5 µl in 28 s - 96-well plate: 10 µl in 5 s, 20 µl in 5 s, 100 µl in 10 s
384-well plate: 1 µl in 6 s, 5 µl in 9 s, 10 µl in 13 s, 20 µl in 21 s
1536-well plate: 1 µl in 15 s, 5 µl in 28 s
- Dispensing accuracy (typical):
- Small tube dispensing cassette: 2 µl, ±10%; 10 µl, ±5%
Standard tube dispensing cassette: 20 µl, ±2%; 100 µl, ±1% - Small tube dispensing cassette: 2 µl, ±10%; 10 µl, ±5%
Standard tube dispensing cassette: 20 µl, ±2%; 100 µl, ±1%
- Dispensing precision:
- Small tube dispensing cassette: 2 μl: CV ≤5%; 10 μl: CV ≤3%
Standard tube dispensing cassette: 20 µl: CV ≤1,5%; 100 μl: CV ≤1% - Small tube dispensing cassette: 2 μl: CV ≤5%; 10 μl: CV ≤3%
Standard tube dispensing cassette: 20 µl: CV ≤1,5%; 100 μl: CV ≤1%
- Dead volume:
- <1 ml for small tube dispensing cassette
<7 ml for standard tube dispensing cassette
Note: Reagents can be backflushed into the original reservoir to avoid loss of expensive reagents. - <1 ml for small tube dispensing cassette
<7 ml for standard tube dispensing cassette
Note: Reagents can be backflushed into the original reservoir to avoid loss of expensive reagents.
- W×D×H:
- 356×328×218 mm
- 356×328×218 mm
- Weight:
- 9,1 kg
- 9,1 kg
Frequently Bought Together