Electroporation Cuvettes Plus
Supplier: BTX
New Product
Certificates
About this item
High quality disposable cuvettes for transfection available in three sizes and compatible with electroporation systems.
- Transfer pipette for manipulation of samples and gamma irradiated together for sterility
- Polished aluminium electrodes in the cuvettes lowers arcing frequency
- Cuvettes (without cap) are autoclavable
Cuvette caps are round for easy, one-handed removal, and are colour-coded for quick identification.