Pipette controller, pipetus®-standard
Supplier: HIRSCHMANN
Certificates
About this item
Pipette controller for pipettes from 0,1 to 100 ml.
- Optimum safety and excellent convenience of use
- Pipettor is lighter and easy to use because the pump is separated from the handle
- Autoclavable, detachable handle for the pipette holder is equipped with two buttons for filling and emptying
This electric pipettor is equipped with a 0,2 µm integrated water-repellent filter (for better durability) and a safety valve to protect the equipment from the introduction of hazardous (contaminated or radioactive) liquids.
Ordering information: The complete standard pipetus® is supplied with an electric pump, two filters and a safety valve.
Related Information
Hirschmann pipetus® HiClass in pipetting
Product Literature
pipetus® standard
Images 360
Frequently Bought Together