Graduated pipettes, type 3, Class B, total delivery
Supplier: HIRSCHMANN
Certificates
About this item
AR-Glas®
- Calibrated to deliver 'Ex', short line graduation
- Marks and inscriptions in amber stain, zero at the top, graduated to the tip
- With cotton plug end
DIN EN ISO 835
Length: 360 mm ±5 mm
Certifications: DIN EN ISO 835
Frequently Bought Together