Magnesium silicate ≥99% (metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Talc, Magnesium silicate, activated, Silicic acid magnesium salt, Florisil
CAS Number:1343-88-0
:
Formula:MgO₃Si
MDL Number:MFCD00078249
Density:3.21
Molecular Weight:100.38 g/mol
Melting Pt:1910 °C
Storage Temperature:Ambient
EINECS:215-681-1
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together