Order Entry
Italy
ContactUsLinkComponent
 
Copper (II) ethylenediamine complex 1 M in water
 
undefined
Copper (II) ethylenediamine complex 1 M in water

 

  • Environmentally Preferable:

 

 

  • Appearance:
    Dark blue Solution
  • Solubility:
    (1.3 g cellulose + 25ml water + 25ml CED complex): clear
  • Ratio:
    (Ethylenediamine to copper) 1.90:1 to 2.10:1
  • Strength:
    0.98 to 1.02 M (Copper molarity)

 

 

Frequently Bought Together