Anti-BET1 Rabbit Polyclonal Antibody
Catalog # PRSI14-775
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:Bet1 golgi vesicular membrane trafficking protein
- Antigen symbol:BET1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Gene ID:10282
- Antigen synonyms:HBET1|Bet1p homolog|blocked early in transport 1 homolog|BET1 homolog|Golgi vesicular membrane trafficking protein p18|golgi vesicular membrane-trafficking protein p18
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at –20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-94 of human BET1 (NP_005859.1)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: BET1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human