Anti-UGDH Rabbit Polyclonal Antibody
PRSI14-260
New Product
- Antibody type:Primary
- Antigen name:UDP-glucose 6-dehydrogenase
- Antigen symbol:UGDH
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Gene ID:7358
- Antigen synonyms:UGDH|UDP-GlcDH|UDP-glucose 6-dehydrogenase|UDPGDH|UDP-Glc dehydrogenase
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at –20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 165-494 of human UGDH (NP_003350.1)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Type: Primary
Antigen: UGDH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat