Anti-USP1 Rabbit Polyclonal Antibody
PRSI22-485
New Product
- Antibody type:Primary
- Antigen name:Ubiquitin carboxyl-terminal hydrolase 1
- Antigen symbol:USP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Gene ID:7398
- Antigen synonyms:Deubiquitinating enzyme 1|USP1|Ubiquitin thioesterase 1|hUBP|Ubiquitin carboxyl-terminal hydrolase 1|Ubiquitin-specific-processing protease 1
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 486-785 of human USP1 (NP_003359.3)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Type: Primary
Antigen: USP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat