Anti-ZNF419 Rabbit Polyclonal Antibody
PRSI59-535
New Product
- Antibody type:Primary
- Antigen name:Zinc finger protein 419
- Antigen symbol:ZNF419
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Epitope:176 - 202 aa
- Form:Liquid
- Gene ID:79744
- Modification:None
- Molecular weight:59 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,5 mg/ml
- Shipping temperature:4°C
- Immunogen:This ZNF419 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 176 - 202 amino acids from the Central region of human ZNF419.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification
- Pk:400 µl
For WB starting dilution is: 1:1000.
Type: Primary
Antigen: ZNF419
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 176-202
Host: Rabbit
Isotype: Ig
Reactivity: Human