Anti-ERV3-1 Rabbit Polyclonal Antibody
PRSI57-042
New Product
- Antibody type:Primary
- Antigen name:Endogenous retrovirus group 3 member 1 Env polyprotein
- Antigen symbol:ERV3-1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Epitope:445-474 aa
- Form:Liquid
- Gene ID:2086
- Modification:None
- Molecular weight:68 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,5 mg/ml
- Shipping temperature:4 °C
- Immunogen:This ERV3 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 445-474 amino acids from the C-terminal region of human ERV3
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification
- Pk:400 µl
For WB starting dilution is: 1:1000 For IHC-P starting dilution is: 1:10~50
Type: Primary
Antigen: ERV3-1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 445-474
Host: Rabbit
Isotype: Ig
Reactivity: Human