Anti-ACPP Rabbit Polyclonal Antibody
Catalog # PRSI18-400
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:Acid phosphatase, prostate
- Antigen symbol:ACPP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Gene ID:55
- Antigen synonyms:Ecto-5'-nucleotidase|5'-NT|Thiamine monophosphatase|ACPP|PAP|Prostatic acid phosphatase|5'-nucleotidase|PAPf39|TMPase
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 40-300 of human ACPP (NP_001090.2)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: ACPP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat