Anti-GLDC Rabbit Polyclonal Antibody
PRSI23-982
New Product
- Antibody type:Primary
- Antigen name:Glycine dehydrogenase (decarboxylating), mitochondrial
- Antigen symbol:GLDC
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Gene ID:2731
- Antigen synonyms:GLDC|Glycine dehydrogenase (aminomethyl-transferring)|GCSP|Glycine decarboxylase|mitochondrial|Glycine dehydrogenase (decarboxylating)|Glycine cleavage system P protein
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 36-290 of human GLDC (NP_000161.2)
- Purification:Affinity purification.
- Pack type:Vial
- Pk:50 µl
Type: Primary
Antigen: GLDC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat