Anti-ADRB2 Recombinant Antibody [Clone: R11/E1]
Catalog # PRSI24-796
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:adrenoceptor beta 2
- Antigen symbol:ADRB2
- Clonality:Recombinant
- Clone:R11/E1
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG kappa
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Form:Liquid
- Gene ID:154
- Antigen synonyms:β-2 adrenoceptor|β2AR|R11E1|Beta-2 adrenergic receptor|Beta-2 adrenoreceptor|Beta-2 adrenoceptor|β-2 adrenergic receptor|β2-AR|β-2 adrenoreceptor
- Modification:Unmodified
- Storage buffer:PBS with 0,02% Proclin 300
- Storage temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping temperature:Shipped on blue ice
- Immunogen:This antibody was raised by immunizing mice with beta-2 adrenergic receptor
- Pack type:Vial
- Pk:0,2 mg
Specifications
About this item
Type: Primary
Antigen: adrenoceptor beta 2
Clonality: Recombinant
Clone: R11/E1
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG kappa
Reactivity: Human