Anti-WDR5 Recombinant Antibody [Clone: RAB-C223]
Catalog # PRSI24-841
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:WD repeat domain 5
- Antigen symbol:WDR5
- Clonality:Recombinant
- Clone:RAB-C223
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:Fab fragment kappa
- Reactivity:
- Environmentally Preferable:
- Cross adsorption:No
- Form:Liquid
- Gene ID:11091
- Antigen synonyms:BMP2-induced 3-kb gene protein|anti-WDR5-RAB-C223|WDR5-A002-03|WD repeat-containing protein 5
- Modification:Unmodified
- Storage buffer:PBS with 0,02% Proclin 300
- Storage temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping temperature:Shipped on blue ice
- Immunogen:This antibody was obtained by recombinant antibody (rAb) phage display recognizing WDR5 protein under non-denaturing conditions
- Pack type:Vial
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: WD repeat domain 5
Clonality: Recombinant
Clone: RAB-C223
Conjugation: Unconjugated
Host: Recombinant
Isotype: Fab fragment kappa
Reactivity: Human