Anti-DDDDK-tag Recombinant Antibody [Clone: M2.1]
PRSI24-503
New Product
- Antibody type:Primary
- Clonality:Recombinant
- Clone:M2.1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Form:Liquid
- Antigen synonyms:AspTyrLysAspAspAspAspLy|DYKDDDDK|Equivalent to FLAG antibodies from Sigma|DDDK|FLAG-tag
- Modification:Unmodified
- Storage buffer:PBS with 0,02% Proclin 300
- Storage temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping temperature:Shipped on blue ice
- Immunogen:Interleukin 2 genetically fused to the peptided DYKDDDDK, corresponding to the DDDDK epitope ta
- Pack type:Vial
- Pk:0,2 mg
Type: Primary
Antigen: DDDDK-tag
Clonality: Recombinant
Clone: M2.1
Conjugation: Unconjugated
Host: Recombinant
Isotype: IgG1 kappa
Reactivity: