3-Isobutyl-1-methylxanthine ≥99% (by HPLC)
Supplier: ENZO LIFE SCIENCES
New Product
Certificates
About this item
Synonyms: 2,6-Dihydroxy-3-isobutyl-1-methylpurine, IBMX, 3,7-Dihydro-3-isobutyl-1-methyl-1H-purine-2,6-dione, 3-Isobutyl-1-methyl-3,7-dihydro-1H-purine-2,6-dione
CAS Number:28822-58-4
:
Formula:C₁₀H₁₄N₄O₂
MDL Number:MFCD00005584
Molecular Weight:222.25 g/mol
Storage Temperature:Freezer
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together