Acetylacetone solution Reag. Ph. Eur. 1000901
Catalog # 87761.180
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:100 mL
- Seq:2440
- Environmentally Preferable:
Specifications
About this item
Synonyms: ACAC, 2,4-Pentanedione
:
:100.12 g/mol
Boiling Pt:140.4
Melting Pt:-23
Flash Pt:30
Storage Temperature:Ambient
EINECS:204-634-0
UN:2310
CAS Number:123-54-6
Formula:CH₃COCH₂COCH₃
MDL Number:MFCD00008787
ADR:3 (6.1) III
Merck Index:13,00083