(Benzyl)triphenylphosphonium chloride ≥99%
: Benzyltriphenylphosphonium chloride
:1100-88-5
:
:C₂₅H₂₂ClP
:MFCD00011913
:1.18 g/cm³ (20 °C)
:388.88 g/mol
:∼ 330 °C (dec.)
:>300 °C (572 °F)
:Ambient
:214-154-3
:3464
:6.1 II
:GHS06
:GHS07
- Environmentally Preferable:
Frequently Bought Together